2-Methylbenzophenone 98%
Supplier: Apollo Scientific
Warning
OR5009-25G OR5009-10G
APOSOR5009-25GEA 24.8 EUR
APOSOR5009-25G APOSOR5009-10G
2-Methylbenzophenone 98%
2-Methylbenzophenone
Formula: C₁₄H₁₂O
Boiling Pt: 309…311 °C
Melting Pt: –18 °C
Density: 1.085 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008518
CAS Number: 131-58-8