3-Bromobenzyl alcohol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Bromophenyl)methanol
OR5020-100G OR5020-500G
APOSOR5020-100GEA 0 EUR
APOSOR5020-100G APOSOR5020-500G
3-Bromobenzyl alcohol 98%
3-Bromobenzyl alcohol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 165 °C/16mmHg
Melting Pt: 110…112 °C
Density: 1.56 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004629
CAS Number: 15852-73-0
EINECS: 239-975-4