3,4-Dichlorobenzyl alcohol 97%
Supplier: Apollo Scientific
Danger
Synonyms: (3,4-Dichlorophenyl)methanol
OR5023-100G OR5023-250G OR5023-50G
APOSOR5023-100GEA 121 EUR
APOSOR5023-100G APOSOR5023-250G APOSOR5023-50G
3,4-Dichlorobenzyl alcohol 97%
3,4-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 125…130 °C/3mmHg
Melting Pt: 36…39 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004633
CAS Number: 1805-32-9
EINECS: 217-299-0
UN: 2811
ADR: 6.1 III