2,4-Dichlorobenzyl alcohol 97%
Supplier: Apollo Scientific
Warning
Synonyms: (2,4-Dichlorophenyl)methanol
OR5024-100G OR5024-25G OR5024-500G
APOSOR5024-100GEA 102 EUR
APOSOR5024-100G APOSOR5024-25G APOSOR5024-500G
2,4-Dichlorobenzyl alcohol 97%
2,4-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 150 °C/25mmHg
Melting Pt: 54…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004606
CAS Number: 1777-82-8
EINECS: 217-210-5
Merck Index: 13,03085