α-Bromo-3-chlorotoulene 98%
Supplier: Apollo Scientific
Danger
Synonyms: α-Bromo-m-chlorotoulene, 3-Chlorobenzyl bromide, 1-(Bromomethyl)-3-chlorobenzene, alpha-Bromo-3-chlorotoluene
OR5035-100G OR5035-25G
APOSOR5035-100GEA 82.8 EUR
APOSOR5035-100G APOSOR5035-25G
α-Bromo-3-chlorotoulene 98%
α-Bromo-3-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 108…110 °C
Melting Pt: 17.5
Density: 1.569 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000597
CAS Number: 766-80-3
EINECS: 212-171-0
UN: 3265
ADR: 8 II