α-Chloro-m-xylene 97%
Supplier: Apollo Scientific
Danger
OR5038-25G OR5038-5G
APOSOR5038-25GEA 61.1 EUR
APOSOR5038-25G APOSOR5038-5G
α-Chloro-m-xylene 97%
α-Chloro-m-xylene
Formula: C₈H₉Cl
Boiling Pt: 195…196 °C
Density: 1.064 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000909
CAS Number: 620-19-9
EINECS: 210-628-9
UN: 3265
ADR: 8,II