3-Acetylpyridine 97+%
Supplier: Apollo Scientific
Danger
Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone
OR5046-100G OR5046-500G OR5046-5G OR5046-25G
APOSOR5046-100GEA 43.5 EUR
APOSOR5046-100G APOSOR5046-500G APOSOR5046-5G APOSOR5046-25G
3-Acetylpyridine 97+%
3-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 218…220 °C
Melting Pt: 41579 °C
Density: 1.102 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1 III
Merck Index: 12,06195