Thiophene-3-carbaldehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde
OR5054-10G OR5054-50G
APOSOR5054-10GEA 21.9 EUR
APOSOR5054-10G APOSOR5054-50G
Thiophene-3-carbaldehyde 98%
Thiophene-3-carbaldehyde
Formula: C₅H₄OS
Boiling Pt: 194…196 °C
Melting Pt: –30 °C
Density: 1.28 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005466
CAS Number: 498-62-4
EINECS: 207-865-5