Ethyl-5-bromonicotinate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 5-bromonicotinate
OR5075-100G OR5075-500G
APOSOR5075-100GEA 109 EUR
APOSOR5075-100G APOSOR5075-500G
Ethyl-5-bromonicotinate 98%
Ethyl 5-bromonicotinate
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Boiling Pt: 86…92 °C/0.5mmHg
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00040366
CAS Number: 20986-40-7