2,6-Dimethoxyphenol 97%
Supplier: Apollo Scientific
Warning
OR5086-25G OR5086-100G
APOSOR5086-25GEA 26.1 EUR
APOSOR5086-25G APOSOR5086-100G
2,6-Dimethoxyphenol 97%
2,6-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 261 °C/760mmHg
Melting Pt: 53…55 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00064434
CAS Number: 91-10-1