1-(Methyldithiocarbonyl)imidazole 95%
Supplier: Apollo Scientific
OR51361-1G OR51361-5G
APOSOR51361-1GEA 87 EUR
APOSOR51361-1G APOSOR51361-5G
1-(Methyldithiocarbonyl)imidazole 95%
1-(Methyldithiocarbonyl)imidazole
Formula: C₅H₆N₂S₂
Melting Pt: 29…33 °C
Density: 1.294 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09750466
CAS Number: 74734-11-5