2'-Methoxyacetophenone 98%
Supplier: Apollo Scientific
Warning
OR5188-100G OR5188-25G
APOSOR5188-100GEA 145 EUR
APOSOR5188-100G APOSOR5188-25G
2'-Methoxyacetophenone 98%
2'-Methoxyacetophenone
Formula: C₉H₁₀O₂
Boiling Pt: 131 °C/18mmHg
Density: 1.09 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00008725
CAS Number: 579-74-8
EINECS: 209-446-2