Dimethyl ether 99%
Supplier: Apollo Scientific
Danger
OR51882-100G OR51882-400G
APOSOR51882-100GEA 482 EUR
APOSOR51882-100G APOSOR51882-400G
Dimethyl ether 99%
Dimethyl ether
Formula: C₂H₆O
Boiling Pt: –24.8 °C
Melting Pt: –141 °C
Density: 0.661 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 115-10-6
EINECS: 204-065-8
UN: 1033
ADR: 2.1,