5-Aminopentan-1-ol 97%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Amino-1-pentanol
OR51934-250G OR51934-25G OR51934-5G OR51934-100G
APOSOR51934-250GEA 0 EUR
APOSOR51934-250G APOSOR51934-25G APOSOR51934-5G APOSOR51934-100G
5-Aminopentan-1-ol 97%
5-Aminopentan-1-ol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 120…122 °C/15.75mmHg
Melting Pt: 33…35 °C
Density: 0.949 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00008237
CAS Number: 2508-29-4
EINECS: 219-718-2
UN: 3259
ADR: 8 III