4-Bromobenzyl mercaptan
Supplier: Apollo Scientific
Warning
Synonyms: p-Bromotoluene-α-thiol
OR51963-1G OR51963-5G
APOSOR51963-1GEA 31.9 EUR
APOSOR51963-1G APOSOR51963-5G
4-Bromobenzyl mercaptan
4-Bromobenzyl mercaptan
Formula: C₇H₇BrS
MW: 203.1
Melting Pt: 26…30 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03701566
CAS Number: 19552-10-4
UN: 2811
ADR: 6.1 III