4-Bromobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: p-Bromobenzoyl chloride
OR52101-100G OR52101-250G OR52101-1KG
APOSOR52101-100GEA 63.8 EUR
APOSOR52101-100G APOSOR52101-250G APOSOR52101-1KG
4-Bromobenzoyl chloride 98%
4-Bromobenzoyl chloride
Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 174 °C/102mmHg
Melting Pt: 36…42 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000683
CAS Number: 586-75-4
EINECS: 209-580-1
UN: 3261
ADR: 8 II