6-Phenylimidazo[2,1-b][1,3]thiazole-5-carbaldehyde
Supplier: Apollo Scientific
Warning
OR52594-1G OR52594-5G
APOSOR52594-1GEA 275 EUR
APOSOR52594-1G APOSOR52594-5G
6-Phenylimidazo[2,1-b][1,3]thiazole-5-carbaldehyde
6-Phenylimidazo[2,1-b][1,3]thiazole-5-carbaldehyde
Formula: C₁₂H₈N₂OS
MW: 228.27 g/mol
Melting Pt: 133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00139592
CAS Number: 74630-73-2