Isovaleronitrile
Supplier: Apollo Scientific
Danger
Synonyms: Isobutyl Cyanide
OR52674-100ML OR52674-25ML OR52674-500ML
APOSOR52674-100MLEA 306 EUR
APOSOR52674-100ML APOSOR52674-25ML APOSOR52674-500ML
Isovaleronitrile
Isovaleronitrile
Formula: C₅H₉N
Boiling Pt: 128…130 °C
Melting Pt: –101 °C
Density: 0.795 g/cm³ (25 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00001944
CAS Number: 625-28-5
EINECS: 210-884-1
UN: 1992
ADR: 3+6.1,III