4-Chlorophenoxyacetyl chloride
Supplier: Apollo Scientific
Danger
OR52691-100G OR52691-25G OR52691-500G
APOSOR52691-100GEA 248 EUR
APOSOR52691-100G APOSOR52691-25G APOSOR52691-500G
4-Chlorophenoxyacetyl chloride
4-Chlorophenoxyacetyl chloride
Formula: C₈H₆Cl₂O₂
Boiling Pt: 165 °C/20mmHg
Melting Pt: 18 °C
Density: 1.37 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000727
CAS Number: 4122-68-3
UN: 3261
ADR: 8,III