Methyl-2,5-dichlorobenzoate 98%
Supplier: Apollo Scientific
Synonyms: Methyl 2,5-dichlorobenzoate
OR52895-100G OR52895-25G
APOSOR52895-100GEA 198 EUR
APOSOR52895-100G APOSOR52895-25G
Methyl-2,5-dichlorobenzoate 98%
Methyl 2,5-dichlorobenzoate
Formula: C₈H₆Cl₂O₂
MW: 205.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000606
CAS Number: 2905-69-3