7,8-Dichloroisoquinoline 98%
Supplier: Apollo Scientific
Warning
OR5292-1G OR5292-250MG OR5292-5G
APOSOR5292-1GEA 140 EUR
APOSOR5292-1G APOSOR5292-250MG APOSOR5292-5G
7,8-Dichloroisoquinoline 98%
7,8-Dichloroisoquinoline
Formula: C₉H₅Cl₂N
Melting Pt: 89…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09258638
CAS Number: 61563-36-8