2-Toluoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Methylbenzoyl chloride, o-Methylbenzoyl chloride, o-Toluoyl chloride, 2-Methylbenzene-1-carbonyl chloride
OR5314-100G OR5314-500G
APOSOR5314-100GEA 65.5 EUR
APOSOR5314-100G APOSOR5314-500G
2-Toluoyl chloride 98%
2-Toluoyl chloride
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 88…90 °C/12mmHg
Melting Pt: -18
Density: 1.187 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00000668
CAS Number: 933-88-0
EINECS: 213-273-8
UN: 3265
ADR: 8 II