Fmoc-trans-4-aminocyclohexane-1-carboxylic acid 99%
Supplier: Apollo Scientific
Synonyms: trans-4-Aminocyclohexanecarboxylic acid, N-FMOC protected
OR53199-1G OR53199-5G
APOSOR53199-1GEA 122 EUR
APOSOR53199-1G APOSOR53199-5G
Fmoc-trans-4-aminocyclohexane-1-carboxylic acid 99%
Fmoc-trans-4-aminocyclohexane-1-carboxylic acid
Formula: C₂₂H₂₃NO₄
MW: 365.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862343
CAS Number: 147900-46-7