3-Hydroxybenzoic acid 99%
Supplier: Apollo Scientific
Warning
Synonyms: m-Hydroxybenzoic Acid
OR5349-100G OR5349-500G OR5349-1KG
APOSOR5349-100GEA 11.8 EUR
APOSOR5349-100G APOSOR5349-500G APOSOR5349-1KG
3-Hydroxybenzoic acid 99%
3-Hydroxybenzoic acid
Formula: C₇H₆O₃
Melting Pt: 200…203 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002506
CAS Number: 99-06-9
EINECS: 202-726-5