4-Amino-6-methoxypyrimidine
Supplier: Apollo Scientific
Warning
OR5359-1G OR5359-10G OR5359-25G
APOSOR5359-1GEA 21.9 EUR
APOSOR5359-1G APOSOR5359-10G APOSOR5359-25G
4-Amino-6-methoxypyrimidine
4-Amino-6-methoxypyrimidine
Formula: C₅H₇N₃O
MW: 125.1286 g/mol
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00129983
CAS Number: 696-45-7