4-Bromobenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
OR5373-100G OR5373-25G OR5373-500G
APOSOR5373-100GEA 94.4 EUR
APOSOR5373-100G APOSOR5373-25G APOSOR5373-500G
4-Bromobenzenesulphonyl chloride
4-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Boiling Pt: 152…154 °C/26mmHg
Melting Pt: 75…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00007437
CAS Number: 98-58-8
EINECS: 202-683-2
UN: 3261
ADR: 8 II
Merck Index: 13,01391