2-Chlorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
OR5374-25G OR5374-100G OR5374-500G
APOSOR5374-25GEA 37.7 EUR
APOSOR5374-25G APOSOR5374-100G APOSOR5374-500G
2-Chlorobenzenesulphonyl chloride 98%
2-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
Boiling Pt: 150…152 °C/12mmHg
Density: 1.548 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00051520
CAS Number: 2905-23-9
EINECS: 220-807-3
UN: 2920
ADR: 8+3,II