2-(Trimethylsilyloxy)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
OR5380-10G OR5380-25G OR5380-50G
APOSOR5380-10GEA 161 EUR
APOSOR5380-10G APOSOR5380-25G APOSOR5380-50G
2-(Trimethylsilyloxy)benzaldehyde 97%
2-(Trimethylsilyloxy)benzaldehyde
Formula: C₁₀H₁₄O₂Si
MW: 194.31 g/mol
Boiling Pt: 116…118 °C
Density: 1.013 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040721
CAS Number: 1078-31-5
EINECS: 214-081-7