1,9-Dibromononane
Supplier: Apollo Scientific
Warning
Synonyms: Nonamethylene dibromide
OR5434-25G OR5434-5G
APOSOR5434-25GEA 24.8 EUR
APOSOR5434-25G APOSOR5434-5G
1,9-Dibromononane
1,9-Dibromononane
Formula: BrH₂C(CH₂)₇CH₂Br
Boiling Pt: 285…288 °C
Melting Pt: –2 °C
Density: 1.407 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000279
CAS Number: 4549-33-1
EINECS: 224-913-0