Phloroglucinol
Supplier: Apollo Scientific
Warning
Synonyms: 1,3,5-Trihydroxybenzene
OR54361-100G OR54361-500G
APOSOR54361-100GEA 94.4 EUR
APOSOR54361-100G APOSOR54361-500G
Phloroglucinol
Phloroglucinol
Formula: C₆H₆O₃
Melting Pt: 215…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002286
CAS Number: 108-73-6
EINECS: 203-611-2