4-(3-Nitrophenyl)morpholine 97+%
Supplier: Apollo Scientific
Warning
OR54369-1G OR54369-25G OR54369-5G
APOSOR54369-1GEA 21.9 EUR
APOSOR54369-1G APOSOR54369-25G APOSOR54369-5G
4-(3-Nitrophenyl)morpholine 97+%
4-(3-Nitrophenyl)morpholine
Formula: C₁₀H₁₂N₂O₃
MW: 208.22 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD03195994
CAS Number: 116922-22-6