4-tert-Butylcalix[4]arene
Supplier: Apollo Scientific
Warning
OR54555-25G OR54555-5G
APOSOR54555-25GEA 191 EUR
APOSOR54555-25G APOSOR54555-5G
4-tert-Butylcalix[4]arene
4-tert-Butylcalix[4]arene
Formula: C₄₄H₅₆O₄
Melting Pt: 342…344 °C
Storage Temperature: Ambient
MDL Number: MFCD00066280
CAS Number: 60705-62-6