(S)-(-)-2-Chlorobutyric acid
Supplier: Apollo Scientific
Danger
OR54621-1G OR54621-5G
APOSOR54621-1GEA 227 EUR
APOSOR54621-1G APOSOR54621-5G
(S)-(-)-2-Chlorobutyric acid
(S)-(-)-2-Chlorobutyric acid
Formula: C₄H₇ClO₂
Boiling Pt: 90…92 °C/12mmHg
Density: 1.19 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00067099
CAS Number: 32653-32-0
UN: 3265
ADR: 8,II