3,3',5,5'-Tetrabromo-2,2'-bithiophene
Supplier: Apollo Scientific
Danger
OR54633-25G OR54633-5G OR54633-1G
APOSOR54633-25GEA 437 EUR
APOSOR54633-25G APOSOR54633-5G APOSOR54633-1G
3,3',5,5'-Tetrabromo-2,2'-bithiophene
3,3',5,5'-Tetrabromo-2,2'-bithiophene
Formula: C₈H₂Br₄S₂
MW: 481.84 g/mol
Melting Pt: 138…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00114807
CAS Number: 125143-53-5