tert-Butyl carbamate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Carbamic acid tert-butyl ester
OR55005-100G OR55005-25G OR55005-500G
APOSOR55005-100GEA 29 EUR
APOSOR55005-100G APOSOR55005-25G APOSOR55005-500G
tert-Butyl carbamate 98%
tert-Butyl carbamate
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007962
CAS Number: 4248-19-5
EINECS: 224-209-3