2,4-Dichloro-3-nitrophenol
Supplier: Apollo Scientific
Warning
OR5510-100MG OR5510-1G OR5510-250MG
APOSOR5510-100MGEA 58.2 EUR
APOSOR5510-100MG APOSOR5510-1G APOSOR5510-250MG
2,4-Dichloro-3-nitrophenol
2,4-Dichloro-3-nitrophenol
Formula: C₆H₃Cl₂NO₃
Melting Pt: 85…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08059572
CAS Number: 38902-87-3
UN: 2811
ADR: 6.1,III