Eicosanedioic acid
Supplier: Apollo Scientific
Warning
OR55106-5G OR55106-10G
APOSOR55106-5GEA 68.3 EUR
APOSOR55106-5G APOSOR55106-10G
Eicosanedioic acid
Eicosanedioic acid
Formula: C₂₀H₃₈O₄
Boiling Pt: 234 °C
Melting Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00059627
CAS Number: 2424-92-2