4-tert-Butylphthalonitrile
Supplier: Apollo Scientific
Synonyms: 4-(tert-Butyl)phthalonitrile, 4-tert-Butyl-1,2-dicyanobenzene
OR55214-25G OR55214-5G
APOSOR55214-25GEA 460 EUR
APOSOR55214-25G APOSOR55214-5G
4-tert-Butylphthalonitrile
4-tert-Butylphthalonitrile
Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Boiling Pt: 165 °C
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00060112
CAS Number: 32703-80-3