[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride 98%
Supplier: Apollo Scientific
Warning
Synonyms: [1,1'-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II), 1,1'-Bis(di-tert-butylphosphino)ferrocene-palladium dichloride, Dichloro[1,1'-bis(di-t-butylphosphino)ferrocene]palladium(II), Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
OR55569-10G OR55569-1G OR55569-5G
APOSOR55569-10GEA 335 EUR
APOSOR55569-10G APOSOR55569-1G APOSOR55569-5G
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride 98%
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride
Formula: C₂₆H₄₄Cl₂FeP₂Pd
MW: 651.75 g/mol
Melting Pt: 203…208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08064219
CAS Number: 95408-45-0