7-Hydroxyflavanone 99%
Supplier: Apollo Scientific
Warning
OR55597-1G OR55597-25G OR55597-5G
APOSOR55597-1GEA 52.2 EUR
APOSOR55597-1G APOSOR55597-25G APOSOR55597-5G
7-Hydroxyflavanone 99%
7-Hydroxyflavanone
Formula: C₁₅H₁₂O₃
Melting Pt: 187…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00017487
CAS Number: 6515-36-2