4-Bromo-1,5-dimethyl-1H-pyrazole-3-carbonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 1,5-Dimethyl-4-bromopyrazole-3-carbonyl chloride
OR55636-1G OR55636-250MG
APOSOR55636-1GEA 357 EUR
APOSOR55636-1G APOSOR55636-250MG
4-Bromo-1,5-dimethyl-1H-pyrazole-3-carbonyl chloride
4-Bromo-1,5-dimethyl-1H-pyrazole-3-carbonyl chloride
Formula: C₆H₆BrClN₂O
Storage Temperature: Ambient
MDL Number: MFCD03421486
CAS Number: 1006471-24-4
UN: 3261
ADR: 8,III