2-Chloro-1-methoxy-3-nitrobenzene 97%
Supplier: Apollo Scientific
Warning
OR55654-1G OR55654-25G OR55654-5G
APOSOR55654-1GEA 21.9 EUR
APOSOR55654-1G APOSOR55654-25G APOSOR55654-5G
2-Chloro-1-methoxy-3-nitrobenzene 97%
2-Chloro-1-methoxy-3-nitrobenzene
Formula: C₇H₆ClNO₃
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD11035883
CAS Number: 3970-39-6