Cyclopentylhydrazine hydrochloride

Supplier: Apollo Scientific
Warning

OR5640-5G OR5640-1G OR5640-10G
APOSOR5640-5GEA 97.3 EUR
APOSOR5640-5G APOSOR5640-1G APOSOR5640-10G
Cyclopentylhydrazine hydrochloride
Cyclopentylhydrazine hydrochloride
Formula: C₅H₁₃ClN₂
Melting Pt: 127…131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320505
CAS Number: 24214-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR