4-(3-Bromophenylsulphonyl)morpholine 95%
Supplier: Apollo Scientific
Warning
OR5722-1G OR5722-5G
APOSOR5722-1GEA 120 EUR
APOSOR5722-1G APOSOR5722-5G
4-(3-Bromophenylsulphonyl)morpholine 95%
4-(3-Bromophenylsulphonyl)morpholine
Formula: C₁₀H₁₂BrNO₃S
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD07363828
CAS Number: 871269-13-5