3-Bromo-N-tert-butylbenzenesulphonamide 98%
Supplier: Apollo Scientific
Synonyms: 3-Bromo-N-(tert-butyl)benzenesulphonamide
OR5730-1G OR5730-5G
APOSOR5730-1GEA 58 EUR
APOSOR5730-1G APOSOR5730-5G
3-Bromo-N-tert-butylbenzenesulphonamide 98%
3-Bromo-N-tert-butylbenzenesulphonamide
Formula: C₁₀H₁₄BrNO₂S
MW: 292.1900 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD07363824
CAS Number: 308283-47-8