2,3-Diaminotoluene 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methylbenzene-1,2-diamine, Toluene-2,3-diamine, 1,2-Diamino-3-methylbenzene
OR5830-1G OR5830-5G
APOSOR5830-1GEA 14.7 EUR
APOSOR5830-1G APOSOR5830-5G
2,3-Diaminotoluene 95%
2,3-Diaminotoluene
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 225 °C
Melting Pt: 59…65 °C
Density: 1.3
Storage Temperature: Ambient
MDL Number: MFCD00011589
CAS Number: 2687-25-4
EINECS: 220-248-5