trans-4-Methylcyclohexylamine
Supplier: Apollo Scientific
Danger
OR59320-100G OR59320-25G OR59320-5G
APOSOR59320-100GEA 231 EUR
APOSOR59320-100G APOSOR59320-25G APOSOR59320-5G
trans-4-Methylcyclohexylamine
trans-4-Methylcyclohexylamine
Formula: C₇H₁₅N
Boiling Pt: 151 °C
Density: 0.86 g/cm³ (25 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD06411232
CAS Number: 2523-55-9
UN: 2734
ADR: 8+3,II