1-Acetyl-6-chloro-1H-indazole
Supplier: Apollo Scientific
Warning
OR59367-1G OR59367-250MG
APOSOR59367-1GEA 176 EUR
APOSOR59367-1G APOSOR59367-250MG
1-Acetyl-6-chloro-1H-indazole
1-Acetyl-6-chloro-1H-indazole
Formula: C₉H₇ClN₂O
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD09878373
CAS Number: 708-40-7