(±)-Indoline-2-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: Indoline-2-carboxylic acid
OR59374-100G OR59374-25G OR59374-5G
APOSOR59374-100GEA 132 EUR
APOSOR59374-100G APOSOR59374-25G APOSOR59374-5G
(±)-Indoline-2-carboxylic acid
(±)-Indoline-2-carboxylic acid
Formula: C₉H₉NO₂
Melting Pt: 168 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010635
CAS Number: 78348-24-0