Azetidine
Supplier: Apollo Scientific
Danger
OR59419-5G OR59419-1G OR59419-25G
APOSOR59419-5GEA 143 EUR
APOSOR59419-5G APOSOR59419-1G APOSOR59419-25G
Azetidine
Azetidine
Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 61…62 °C
Density: 0.847 g/cm³ (25 °C)
Flash Pt: –21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005165
CAS Number: 503-29-7
EINECS: 207-963-8
UN: 2733
ADR: 3+8 II